accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONH(S)OCl

CAS RN 402-71-1

IUPAC Name: N-[(2S)-4-chloro-3-oxo-1-phenylbutan-2-yl]-4-methylbenzene-1-sulfonamide
Synonyms: TPCK C17H18ClNO3S tosylphenylalanyl chloromethyl ketone tosyl phenylalanyl chloromethyl ketone chloromethane, tosylphenylalanyl ketone, tosylphenylalanyl chloromethyl tosylphenylalanyl chloromethane chloromethyl ketone, tosylphenylalanyl
Molecular Weight (g/mol): 351.85
Molecular Formula: C17H18ClNO3S
InChi Key: MQUQNUAYKLCRME-INIZCTEOSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CC1=CC=CC=C1)C(=O)CCl
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results