Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(p-Toluenesulfonyl)-L-phenylalanyl Chloromethyl Ketone 95.0+%, TCI America™
SDP

Catalog No. T28101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28101G Supplier TCI America Supplier No. T28101G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 402-71-1
Molecular Formula C17H18ClNO3S
Molecular Weight (g/mol) 351.85
MDL Number MFCD00000935
InChI Key MQUQNUAYKLCRME-INIZCTEOSA-N
Synonym tpck, tos-phe-ch2cl, tosylphenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanine chloromethyl ketone, n-p-tosyl-l-phenylalanine chloromethyl ketone, l-1-tosylamido-2-phenylethyl chloromethyl ketone, l-n-alpha-chloroacetyl phenethyl-p-toluenesulfonamide, tos-phe-chloromethylketone, chembl60718
PubChem CID 439647
ChEBI CHEBI:9642
IUPAC Name N-[(2S)-4-chloro-3-oxo-1-phenylbutan-2-yl]-4-methylbenzene-1-sulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CC1=CC=CC=C1)C(=O)CCl

Specifications

Melting Point 108°C
Color White
Quantity 1 g
Formula Weight 351.85
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(p-Toluenesulfonyl)-L-phenylalanyl Chloromethyl Ketone
RTECSNumber XT5613500
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.