accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOO

CAS RN 40204-26-0

IUPAC Name: 3-(benzyloxy)-3-oxopropanoic acid
Synonyms: 3-(benzyloxy)-3-oxopropanoic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: CFLAHQSWDKNWPW-UHFFFAOYSA-N
SMILES: OC(=O)CC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results