accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40228-90-8

IUPAC Name: 7-phenylheptanoic acid
Synonyms: 7-phenylheptanoic acid
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: ZVSXKFNTWOIGJI-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results