Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 7-Phenylheptanoic acid

Catalog No. CDS000796 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS000796 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS000796 Supplier Sigma Aldrich Supplier No. CDS000796100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,8-dichlorooctane is a chloroparaffin. the non-exponentiality of proton spin-lattice relaxation in polycrystalline 1,8-dichlorooctane in zeeman-quadrupole level-crossing experiments at various frequencies is reported. it has important synthetic applications in food, pharmaceutical, biomedical, electrical and electronic sectors.1,8-dichlorooctane may be used:
  • As adsorbate to investigate the performance of carbonaceous materials for the adsorption of emerging pollutants
  • To prepare the c20-backbone of 1,20-substituted eicosanes carrying phosphate headgroups and readily derivatizable thiol, maleimido, and activated carboxylic ester moieties
  • In the synthesis of planar-chiral imidazolium salt
  • In the preparation of biosensors
  • In the fabrication of strong polymeric membranes for concentrating amino acids, having applications in food and pharmaceutical industry
  • In the synthesis of perfluorocarbons having biomedical applications

Specifications

CAS 40228-90-8
Molecular Formula C13H18O2
Linear Formula C13H18O2
MDL Number MFCD00037801
Quantity 100 g
Molecular Weight (g/mol) 206.28
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.