accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40241-78-9

IUPAC Name: hydrogen 2-(methylsulfanyl)-4,5-dihydro-1H-imidazole bromide
Synonyms: hydrogen 2-(methylsulfanyl)-4,5-dihydro-1H-imidazole bromide
Molecular Weight (g/mol): 197.09
Molecular Formula: C4H9BrN2S
InChi Key: UPISCLUJZRQPPJ-UHFFFAOYSA-N
SMILES: [H+].[Br-].CSC1=NCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results