Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)-4,5-dihydro-1H-imidazole hydrobromide, Thermo Scientific™

Catalog No. DP01481EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
DP01481EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DP01481EA Supplier Thermo Scientific Chemicals Supplier No. DP01481EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40241-78-9
Molecular Formula C4H9BrN2S
Molecular Weight (g/mol) 197.094
MDL Number MFCD01570110
InChI Key UPISCLUJZRQPPJ-UHFFFAOYSA-N
Synonym 2-methylthio-4,5-dihydro-1h-imidazole hydrobromide, 4,5-dihydro-2-methylthio-1h-imidazole monohydrobromide, 2-methylsulfanyl-4,5-dihydro-1h-imidazole hydrobromide, 2-methylthio-2-imidazoline, bromide, 1h-imidazole,4,5-dihydro-2-methylthio-, hydrobromide 1:1
PubChem CID 2777335
IUPAC Name 2-methylsulfanyl-4,5-dihydro-1H-imidazole;hydrobromide
SMILES CSC1=NCCN1.Br

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 197.1
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(Methylthio)-4, 5-dihydro-1H-imidazole hydrobromide
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.