accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4028-23-3

IUPAC Name: chlorodimethyl(prop-2-en-1-yl)silane
Synonyms: allyl-chloro-dimethyl silane
Molecular Weight (g/mol): 134.68
Molecular Formula: C5H11ClSi
InChi Key: KMVZWUQHMJAWSY-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CC=C
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)