accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 402822-72-4

IUPAC Name: {2'-methyl-[1,1'-biphenyl]-2-yl}diphenylphosphane
Synonyms: {2'-methyl-[1,1'-biphenyl]-2-yl}diphenylphosphane
Molecular Weight (g/mol): 352.42
Molecular Formula: C25H21P
InChi Key: GUENDLLDUNRPIC-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results