CAS RN 402832-01-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Caspase-4 Inhibitor I
The Caspase-4 Inhibitor I, also referenced under CAS 402832-01-3, controls the biological activity of Caspase-4. This small molecule/inhibitor is primarily used for Cancer applications.