Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-4 Inhibitor I

Catalog No. 2187551MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-875-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-875-51MG Supplier MilliporeSigma™ Supplier No. 2187551MG
Add to Cart
Add to Cart

The Caspase-4 Inhibitor I, also referenced under CAS 402832-01-3, controls the biological activity of Caspase-4. This small molecule/inhibitor is primarily used for Cancer applications.

Peptide Sequence: Ac-Leu-Glu-Val-Asp-CHO
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins caspase-4
CAS 402832-01-3
Molecular Formula C22H36N4O9
Formula Weight 500.6g/mol
Solubility DMSO or H2O
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.