CAS RN 402957-28-2
IUPAC Name:
3-{[(1S,3aR,6aS)-2-[(2S)-2-[(2S)-2-cyclohexyl-2-[(pyrazin-2-yl)formamido]acetamido]-3,3-dimethylbutanoyl]-octahydrocyclopenta[c]pyrrol-1-yl]formamido}-N-cyclopropyl-2-oxohexanamide
Synonyms:
telaprevir
Molecular Weight (g/mol):
679.86
Molecular Formula:
C36H53N7O6
InChi Key:
BBAWEDCPNXPBQM-IGSHNTALSA-N
SMILES:
CCCC(NC(=O)[C@@H]1[C@H]2CCC[C@H]2CN1C(=O)[C@@H](NC(=O)[C@@H](NC(=O)C1=CN=CC=N1)C1CCCCC1)C(C)(C)C)C(=O)C(=O)NC1CC1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Telaprevir
Thermo Scientific Chemicals Telaprevir
CAS: 402957-28-2 Molecular Formula: C36H53N7O6