accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClF

CAS RN 403-17-8

IUPAC Name: 4-chloro-3-fluorobenzoic acid
Synonyms: 4-chloro-3-fluorobenzoic acid
Molecular Weight (g/mol): 174.56
Molecular Formula: C7H4ClFO2
InChi Key: QPIBHIXKUQKNFP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C(F)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results