Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-fluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C28915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C28915G Supplier TCI America Supplier No. C28915G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 403-17-8
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.56
MDL Number MFCD00143290
InChI Key QPIBHIXKUQKNFP-UHFFFAOYSA-N
Synonym 3-fluoro-4-chlorobenzoic acid, 4-chloro-3-fluorobenzoicacid, 4-chloro-3-fluoro-benzoic acid, benzoic acid, 4-chloro-3-fluoro, pubchem1333, acmc-1ctvr, ksc493s7r, 4-chloro-3-fluoro benzoicacid, rarechem al bo 1770, 4-chloro-3-fluoro benzoic acid
PubChem CID 2736536
IUPAC Name 4-chloro-3-fluorobenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C(F)=C1

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.