accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOFNOO

CAS RN 403-21-4

IUPAC Name: 3-fluoro-4-nitrobenzoic acid
Synonyms: 3-fluoro-4-nitrobenzoic acid
Molecular Weight (g/mol): 185.11
Molecular Formula: C7H4FNO4
InChi Key: WVZBIQSKLXJFNX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C(F)=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)