accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONH2OHHOF

CAS RN 403-90-7

IUPAC Name: 2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
Synonyms: pardinon|m-ftyr|fluorthyrin
Molecular Weight (g/mol): 199.18
Molecular Formula: C9H10FNO3
InChi Key: VIIAUOZUUGXERI-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(O)C(F)=C1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)