Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-DL-tyrosine, 97%, Thermo Scientific Chemicals

Catalog No. AAL01479ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL01479ME 500 mg
AAL01479MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL01479ME Supplier Thermo Scientific Chemicals Supplier No. L01479ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.

Solubility
Soluble in water (28 g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-90-7
Molecular Formula C9H10FNO3
Molecular Weight (g/mol) 199.18
MDL Number MFCD00063075
InChI Key VIIAUOZUUGXERI-UHFFFAOYNA-N
Synonym 3-fluoro-dl-tyrosine, 2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, pardinon, m-fluoro-dl-tyrosine, fluorthyrin, 3-fluorotyrosin, 3-fluorotyrosine, m-ftyr, dl-m-fluorotyrosine, dl-3-fluorotyrosine
PubChem CID 92100
ChEBI CHEBI:32771
IUPAC Name 2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=C(O)C(F)=C1)C(O)=O

Specifications

Melting Point ∽280°C (decomposition)
Beilstein 3204804
Solubility Information Soluble in water (28g/L).
Quantity 500 mg
Formula Weight 199.18
Percent Purity 98%
Chemical Name or Material 3-Fluoro-DL-tyrosine
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 206-964-0
RTECSNumber YP2660000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.