accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO

CAS RN 4036-30-0

IUPAC Name: 2-phenylbutanedioic acid
Synonyms: 2-phenylbutanedioic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: LVFFZQQWIZURIO-UHFFFAOYNA-N
SMILES: OC(=O)CC(C(O)=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results