Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Phenylsuccinic Acid 98.0+%, TCI America™
SDP

Catalog No. P09361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P09361G Supplier TCI America Supplier No. P09361G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4036-30-0
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00065929
InChI Key LVFFZQQWIZURIO-QMMMGPOBSA-N
Synonym s-+-phenylsuccinic acid, s-2-phenylsuccinic acid, s-2-phenyl succinic acid, 2s-2-phenylbutanedioic acid, butanedioic acid, phenyl-, 2s, 2s-2-phenylbutanedioic acid ee, s-phenylsuccinic acid, 2s-2-phenylsuccinic acid, s-+-phenylsuccinicacid, qspl 009
PubChem CID 736146
IUPAC Name (2S)-2-phenylbutanedioic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)C(=O)O

Specifications

Melting Point 174°C
Color White
Quantity 1 g
Formula Weight 194.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-Phenylsuccinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.