accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrClBr

CAS RN 40360-47-2

IUPAC Name: 3,5-dibromo-2-chloropyridine
Synonyms: 3,5-dibromo-2-chloropyridine
Molecular Weight (g/mol): 271.34
Molecular Formula: C5H2Br2ClN
InChi Key: PYSICVOJSJMFKP-UHFFFAOYSA-N
SMILES: ClC1=C(Br)C=C(Br)C=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results