accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOONOO

CAS RN 40392-27-6

IUPAC Name: 5-nitro-2-phenyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 5-nitro-2-phenylisoindole-1,3-dione
Molecular Weight (g/mol): 268.23
Molecular Formula: C14H8N2O4
InChi Key: HNNCFMUZZXJQLR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C(C=C1)C(=O)N(C2=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results