Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-N-phenylphthalimide 98.0+%, TCI America™
SDP

Catalog No. N057425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N057425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N057425G Supplier TCI America Supplier No. N057425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40392-27-6
Molecular Formula C14H8N2O4
Molecular Weight (g/mol) 268.228
MDL Number MFCD00191559
InChI Key HNNCFMUZZXJQLR-UHFFFAOYSA-N
PubChem CID 762600
IUPAC Name 5-nitro-2-phenylisoindole-1,3-dione
SMILES C1=CC=C(C=C1)N2C(=O)C3=C(C2=O)C=C(C=C3)[N+](=O)[O-]

Specifications

Melting Point 187°C
Color Yellow
Quantity 25 g
Formula Weight 268.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-N-phenylphthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.