accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40398-01-4

IUPAC Name: 1-chloro-2-isocyanato-3-methylbenzene
Synonyms: 1-chloro-2-isocyanato-3-methylbenzene
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: FBTQQNYGMICJQZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Cl)=C1N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results