accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40412-06-4

H3CSOOOS

CAS RN 40412-06-4

IUPAC Name: 2-(thiophen-2-yl)ethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-(thiophen-2-yl)ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 282.37
Molecular Formula: C13H14O3S2
InChi Key: HLPRKWVEMYDPAU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCC1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results