accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3SiH3CCH3ON(E/Z)

CAS RN 404392-70-7

IUPAC Name: tert-butyldimethylsilyl N-phenylbenzenecarboximidate
Synonyms: tert-butyldimethylsilyl N-phenylbenzenecarboximidate
Molecular Weight (g/mol): 311.5
Molecular Formula: C19H25NOSi
InChi Key: CQKWSHDUCPVOAI-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC(=NC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results