CAS RN 40493-18-3
CAS RN 40493-18-3
IUPAC Name:
3-chloro-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6-tetraene
Synonyms:
3-chloro-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6-tetraene
Molecular Weight (g/mol):
224.71
Molecular Formula:
C10H9ClN2S
InChi Key:
PRNJDUCSVXTOTN-UHFFFAOYSA-N
SMILES:
ClC1=C2C3=C(CCCC3)SC2=NC=N1
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 4-Chloro-5H,6H,7H,8H-pyrimido[4,5-b]benzothiophene
Sigma Aldrich 4-Chloro-5H,6H,7H,8H-pyrimido[4,5-b]benzothiophene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.