CAS RN 405-39-0
IUPAC Name:
2,6-di{[(benzyloxy)carbonyl]amino}hexanoic acid
Synonyms:
2,6-di{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol):
414.46
Molecular Formula:
C22H26N2O6
InChi Key:
BLZXFNUZFTZCFD-UHFFFAOYNA-N
SMILES:
OC(=O)C(CCCCNC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
Z-Lys Z , Advanced ChemTech
CAS: 405-39-0 Molecular Formula: C22H26N2O6 Molecular Weight (g/mol): 414.46 MDL Number: MFCD00037822 InChI Key: BLZXFNUZFTZCFD-UHFFFAOYNA-N IUPAC Name: 2,6-di{[(benzyloxy)carbonyl]amino}hexanoic acid SMILES: OC(=O)C(CCCCNC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1