accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFSOOFFFSOOFFF

CAS RN 405074-81-9

IUPAC Name: 1-(ditrifluoromethanesulfonylmethyl)-2,3,4,5,6-pentafluorobenzene
Synonyms: 1-(ditrifluoromethanesulfonylmethyl)-2,3,4,5,6-pentafluorobenzene
Molecular Weight (g/mol): 446.21
Molecular Formula: C9HF11O4S2
InChi Key: RLLDXJXYMKTGPV-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results