accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrClN

CAS RN 405224-23-9

IUPAC Name: 5-bromo-2-chloropyridine-3-carbonitrile
Synonyms: 5-bromo-2-chloropyridine-3-carbonitrile
Molecular Weight (g/mol): 217.45
Molecular Formula: C6H2BrClN2
InChi Key: MQOHJAYYYVQBSH-UHFFFAOYSA-N
SMILES: ClC1=NC=C(Br)C=C1C#N
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)