Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chloro-3-cyanopyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAH6166406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6166406 5 g
AAH6166403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6166406 Supplier Thermo Scientific Chemicals Supplier No. H6166406

5-Bromo-2-chloro-3-cyanopyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chloro-3-cyanopyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405224-23-9
Molecular Formula C6H2BrClN2
Molecular Weight (g/mol) 217.45
MDL Number MFCD09801046
InChI Key MQOHJAYYYVQBSH-UHFFFAOYSA-N
Synonym 5-bromo-2-chloronicotinonitrile, 5-bromo-2-chloro-3-cyanopyridine, 5-bromo-2-chloro-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 5-bromo-2-chloro, pubchem15627, acmc-1amn3, 5-bromo-2-chloronicotinontrile, 5-bromo-2-chloro-nicotinonitrile, 5-bromo-2-chloro-pyridine-3-carbonitrile, 5-bromo-2-chloropyridine-3-carbonitrile
PubChem CID 21948269
IUPAC Name 5-bromo-2-chloropyridine-3-carbonitrile
SMILES ClC1=NC=C(Br)C=C1C#N

Specifications

Quantity 5 g
UN Number UN3439
Solubility Information Slightly soluble in water.
Formula Weight 217.45
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-chloro-3-cyanopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.