accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 405225-21-0

IUPAC Name: 2-[(3-{[1,1'-biphenyl]-4-yloxy}-3-(4-fluorophenyl)propyl)(methyl)amino]acetic acid
Synonyms: [(3-{[1,1'-biphenyl]-4-yloxy}-3-(4-fluorophenyl)propyl)(methyl)amino]acetic acid
Molecular Weight (g/mol): 393.46
Molecular Formula: C24H24FNO3
InChi Key: FDORQEIHOKEJNX-UHFFFAOYNA-N
SMILES: CN(CCC(OC1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(F)C=C1)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results