accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40530-18-5

IUPAC Name: 5-bromo-2-hydroxybenzonitrile
Synonyms: 5-bromo-2-hydroxybenzonitrile
Molecular Weight (g/mol): 198.02
Molecular Formula: C7H4BrNO
InChi Key: PVCONXMDUZOPJH-UHFFFAOYSA-N
SMILES: OC1=CC=C(Br)C=C1C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results