accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOH2NONOOFFF

CAS RN 40533-25-3

IUPAC Name: 2-carbamoyl-7-nitro-5-(trifluoromethyl)-1,3-benzothiazol-3-ium-3-olate
Synonyms: 2-carbamoyl-7-nitro-5-(trifluoromethyl)-1,3-benzothiazol-3-ium-3-olate
Molecular Weight (g/mol): 307.2
Molecular Formula: C9H4F3N3O4S
InChi Key: FHFLXKUKCSDMLD-UHFFFAOYSA-N
SMILES: NC(=O)C1=[N+]([O-])C2=CC(=CC(=C2S1)[N+]([O-])=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results