accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 40546-94-9

IUPAC Name: 6-methyl-4-phenyl-3,4-dihydro-2H-1-benzopyran-2-one
Synonyms: 6-methyl-4-phenyl-3,4-dihydro-1-benzopyran-2-one
Molecular Weight (g/mol): 238.29
Molecular Formula: C16H14O2
InChi Key: SUHIZPDCJOQZLN-UHFFFAOYNA-N
SMILES: CC1=CC2=C(OC(=O)CC2C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results