Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-4-phenyl-2-chromanone 98.0+%, TCI America™
SDP

Catalog No. M20935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20935G 5 g
M209325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20935G Supplier TCI America Supplier No. M20935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40546-94-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.29
MDL Number MFCD03695019
InChI Key SUHIZPDCJOQZLN-UHFFFAOYNA-N
Synonym 3,4-Dihydro-6-methyl-4-phenylcoumarin
PubChem CID 3791643
IUPAC Name 6-methyl-4-phenyl-3,4-dihydro-2H-1-benzopyran-2-one
SMILES CC1=CC2=C(OC(=O)CC2C2=CC=CC=C2)C=C1

Specifications

Melting Point 82°C
Color White
Boiling Point 230°C
Quantity 5 g
Formula Weight 238.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-4-phenyl-2-chromanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.