accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFOOOOOFFFFFFFFFFFFFF

CAS RN 40573-09-9

IUPAC Name: hexadecafluoro-2,4,6,8,11-pentaoxatridec-12-ene
Synonyms: hexadecafluoro-2,4,6,8,11-pentaoxatridec-12-ene
Molecular Weight (g/mol): 480.06
Molecular Formula: C8F16O5
InChi Key: HCGYVDVMTQESCQ-UHFFFAOYSA-N
SMILES: FC(F)=C(F)OC(F)(F)C(F)(F)OC(F)(F)OC(F)(F)OC(F)(F)OC(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)