CAS RN 40575-42-6
IUPAC Name:
(4R)-oct-1-en-4-ol
Synonyms:
(4R)-oct-1-en-4-ol
Molecular Weight (g/mol):
128.22
Molecular Formula:
C8H16O
InChi Key:
UZGCMRVEDHLBGY-QMMMGPOBSA-N
SMILES:
CCCC[C@@H](O)CC=C
1
–
1
of
1
results
1
Promotions Available
1-Octen-4-ol, 99%
1-Octen-4-ol, 99%
CAS: 40575-42-6 Molecular Formula: C8H16O Molecular Weight (g/mol): 128.22 MDL Number: MFCD00021943 InChI Key: UZGCMRVEDHLBGY-QMMMGPOBSA-N Synonym: 1-octen-4-ol,2e-octen-4-ol,acmc-1akyk PubChem CID: 142471 IUPAC Name: oct-1-en-4-ol SMILES: CCCC[C@@H](O)CC=C