Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octen-4-ol, 99%

Catalog No. AAB2070714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2070714 25 g
AAB2070706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2070714 Supplier Thermo Scientific Chemicals Supplier No. B2070714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 40575-42-6 | C8H16O | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40575-42-6
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00021943
InChI Key UZGCMRVEDHLBGY-QMMMGPOBSA-N
Synonym 1-octen-4-ol, 2e-octen-4-ol, acmc-1akyk
PubChem CID 142471
IUPAC Name oct-1-en-4-ol
SMILES CCCC[C@@H](O)CC=C

Specifications

Density 0.83
Quantity 25 g
Formula Weight 128.22
Percent Purity 99%
Chemical Name or Material 1-Octen-4-ol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.