accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNCl

CAS RN 405930-65-6

IUPAC Name: 4-(benzyloxy)-6-chloropyrimidine
Synonyms: 4-(benzyloxy)-6-chloropyrimidine
Molecular Weight (g/mol): 220.66
Molecular Formula: C11H9ClN2O
InChi Key: KXDXFVQZZGSEBU-UHFFFAOYSA-N
SMILES: ClC1=NC=NC(OCC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results