CAS RN 406-78-0
CAS RN 406-78-0
IUPAC Name:
1,1,2,2-tetrafluoro-1-(2,2,2-trifluoroethoxy)ethane
Synonyms:
1,1,2,2-tetrafluoro-1-(2,2,2-trifluoroethoxy)ethane
Molecular Weight (g/mol):
200.06
Molecular Formula:
C4H3F7O
InChi Key:
CWIFAKBLLXGZIC-UHFFFAOYSA-N
SMILES:
FC(F)C(F)(F)OCC(F)(F)F
1
–
1
of
1
results
1
1,1,2,2-Tetrafluoroethyl 2,2,2-Trifluoroethyl Ether 99.0+%, TCI America™
CAS: 406-78-0 Molecular Formula: C4H3F7O Molecular Weight (g/mol): 200.056 MDL Number: MFCD00236118 InChI Key: CWIFAKBLLXGZIC-UHFFFAOYSA-N PubChem CID: 164596 IUPAC Name: 1,1,2,2-tetrafluoro-1-(2,2,2-trifluoroethoxy)ethane SMILES: C(C(F)(F)F)OC(C(F)F)(F)F