accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNCH3

CAS RN 40611-85-6

IUPAC Name: ethyl 4-methyl-1H-pyrrole-2-carboxylate
Synonyms: ethyl 4-methyl-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: RWFKYBVNHRKZSN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(C)=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results