Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Methyl-2-pyrrolecarboxylate 97.0+%, TCI America™
SDP

Catalog No. M11455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11455G 5 g
M11451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11455G Supplier TCI America Supplier No. M11455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40611-85-6
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00191668
InChI Key RWFKYBVNHRKZSN-UHFFFAOYSA-N
Synonym 4-Methyl-2-pyrrolecarboxylic Acid Ethyl Ester
PubChem CID 643361
IUPAC Name ethyl 4-methyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=CC(C)=CN1

Specifications

Melting Point 43°C
Color White
Boiling Point 104°C
Quantity 5 g
Formula Weight 153.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Methyl-2-pyrrolecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.