accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4062-60-6

IUPAC Name: tert-butyl[2-(tert-butylamino)ethyl]amine
Synonyms: tert-butyl[2-(tert-butylamino)ethyl]amine
Molecular Weight (g/mol): 172.32
Molecular Formula: C10H24N2
InChi Key: KGHYGBGIWLNFAV-UHFFFAOYSA-N
SMILES: CC(C)(C)NCCNC(C)(C)C
Boiling Point 
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)