accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 406463-06-7

IUPAC Name: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Synonyms: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Weight (g/mol): 255.12
Molecular Formula: C15H18BNO2
InChi Key: VMFALDWCEQUFSX-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=CC=CN=C2C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)