Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorodicyclopentylphosphine

Catalog No. 6490665G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-906-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 64-906-65G Supplier Sigma Aldrich Supplier No. 6490665G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorodicyclopentylphosphine may be used to prepare:Cp,Cp-MannOP, a chiral diphosphinite ligand by reacting with mannitol.[Cp=cyclopentyl; Mann=mannitol](S)-Cp,Cp-oxoProNOP, a bis(dicycloalkylphosphino) amidophosphine-phosphinite ligand by reacting with (S)-5-(hydroxymethyl)-2-pyrrolidinoneLcPeNiCl, a precursor for preparing LcPeNiH, which is an efficient catalyst for the reduction of CO2[cPe=cyclopentyl]

Specifications

CAS 406463-06-7
Melting Point 62°C to 66°C
Molecular Formula C15H18BNO2
Linear Formula C15H18BNO2
MDL Number MFCD06411327
Quantity 5 g
Synonym Dicyclopentylchlorophosphine; Dicyclopentylphosphine chloride
Molecular Weight (g/mol) 255.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.