accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40648-92-8

IUPAC Name: bis[(1S)-1-phenylethyl]azanium
Synonyms: bis[(1S)-1-phenylethyl]azanium
Molecular Weight (g/mol): 226.34
Molecular Formula: C16H20N
InChi Key: NXLACVVNHYIYJN-KBPBESRZSA-O
SMILES: C[C@H]([NH2+][C@@H](C)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)