Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-(-)-Bis(alpha-methylbenzyl)amine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B20355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B20355G Supplier TCI America Supplier No. B20355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40648-92-8
Molecular Formula C16H20ClN
Molecular Weight (g/mol) 261.793
MDL Number MFCD00216671
InChI Key ZBQCLJZOKDRAOW-IODNYQNNSA-N
Synonym s-bis s-1-phenylethyl amine hydrochloride, s,s---bis alpha-methylbenzyl amine hydrochloride, --bis s-1-phenylethyl amine hydrochloride, s,s---bis a-methylbenzyl amine hydrochloride, s,s---bis 1-phenylethyl amine hydrochloride, bis 1s-1-phenylethyl amine hydrochloride, s,s---bis alpha-methylbenzyl aminehydrochloride, pubchem8138, --bis s-, a-methylbenzyl amine hydrochloride, 1s-1-phenyl-n-1s-1-phenylethyl ethanamine;hydrochloride
PubChem CID 12353077
IUPAC Name (1S)-1-phenyl-N-[(1S)-1-phenylethyl]ethanamine;hydrochloride
SMILES CC(C1=CC=CC=C1)NC(C)C2=CC=CC=C2.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 261.79
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-(-)-Bis(alpha-methylbenzyl)amine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.