accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 406482-20-0

IUPAC Name: (2,6-difluoro-4-methoxyphenyl)boronic acid
Synonyms: 2,6-difluoro-4-methoxyphenylboronic acid
Molecular Weight (g/mol): 187.94
Molecular Formula: C7H7BF2O3
InChi Key: WVSZSFADEBGONQ-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(B(O)O)C(F)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results