Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D47281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D47281G Supplier TCI America Supplier No. D47281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 406482-20-0
Molecular Formula C7H7BF2O3
Molecular Weight (g/mol) 187.94
MDL Number MFCD05664299
InChI Key WVSZSFADEBGONQ-UHFFFAOYSA-N
Synonym 2,6-Difluoro-4-methoxybenzeneboronic Acid
PubChem CID 16217424
IUPAC Name (2,6-difluoro-4-methoxyphenyl)boronic acid
SMILES COC1=CC(F)=C(B(O)O)C(F)=C1

Specifications

Melting Point 130°C
Color White
Quantity 1 g
Formula Weight 187.94
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluoro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.