accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4066-02-8

IUPAC Name: 2-cyclohexyl-6-[(3-cyclohexyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Synonyms: 2-cyclohexyl-6-[(3-cyclohexyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Molecular Weight (g/mol): 392.58
Molecular Formula: C27H36O2
InChi Key: AKNMPWVTPUHKCG-UHFFFAOYSA-N
SMILES: CC1=CC(CC2=CC(C)=CC(C3CCCCC3)=C2O)=C(O)C(=C1)C1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results